PIG® Overpacks
Leverandør: NEW PIG
PAK708-WH PAK709-WH PAK725-WH
NEWPPAK708-WHEA 2800 NOK
NEWPPAK708-WH NEWPPAK709-WH NEWPPAK725-WH
PIG® Overpacks
Tønner, fat, trommer
Tough enough for rough handling, guaranteed. Ship your haz waste in our UN rated overpacks to stay compliant with regs.
  • Containers are nestable when empty to save storage space
  • Designed and proven to withstand the abuse of shipping to help keep hazardous waste safely contained
  • Material with UV inhibitors is lightweight and chemical-resistant
X-rated for packing groups I, II and III, so it's ideal for regulated packaging applications, including shipping hazardous waste by land, sea or rail.
NB: If using this product with flammable liquids, pleaseMore Product Information