Soil sieves
Leverandør: NICKEL ELECTRO
9130
NICK9130EA 4050 NOK
NICK9130
Soil sieves
Sikter Sikter, test
These soil sieves designed for educational use have an aluminium body with stainless steel mesh.
This complete set of soil sieves consists of nine sieve sizes with a lid and pan.