Swabs for culture media, ready-to-use, Compact Dry
Leverandør: Shimadzu Diagnostics Europe
450001NRS-0050 450003LTE-0050 450004BPW-0050
NISS450001NRS-0050EA 1150 NOK
NISS450001NRS-0050 NISS450003LTE-0050 NISS450004BPW-0050
Swabs for culture media, ready-to-use, Compact Dry
Svabere Svabere for klinisk bruk
CompactDry Swabs for surfaces, meat, fish and poultry. New design with swabs pre immersed in 1 ml buffer, ready for use with a self-standing tube and a high visibility blue cap and swab.
NRS (Neutralising Rinse Solution): Universal neutralising solution suitable for testing most disinfected surfaces within the food, cosmetic and pharmaceutical industries.
PBS (Phosphate Buffered Saline): Designed for environmental/surface sample collection for the food and beverage industry.
LTE (Letheen Broth): Used to sample surfaces treated with a quaternary ammonium-based disinfectants (More Product Information