Cleanroom face masks, BioClean DB™, PE
Leverandør: Nitritex
BDBN-G
NITRBDBN-GEA 5800 NOK
NITRBDBN-G
Cleanroom face masks, BioClean DB™, PE
Masker Munnbind, renrom
The non-sterile pouch style mask has knitted headbands and all seams are ultrasonically bonded. The mask has inner and outer facing layers constructed of spunbond non woven polyester with the middle filter layer made from meltblown polyester.
  • Integral neck guard
  • Latex-free
  • Patented spacer maintains clear breathing space
  • Ultrasonically sealed edges
  • Low-linting
  • Fully enclosed malleable nose-band
Suitable for use in ISO class 4, cleanroom environments.
Store in a dry, cool place (<40 °C) away from direct sunlight and fluorescent light.