MEA cleanroom looped face mask, BioClean™
Leverandør: Nitritex
MEA210-2
NITRMEA210-2EA 10675 NOK
NITRMEA210-2
MEA cleanroom looped face mask, BioClean™
Masker Munnbind, renrom
Non sterile ear loop face mask.
  • High bacterial, viral, and particle efficiency filtration
  • Fully enclosed malleable noseband
  • Ultrasonically sealed edges
  • Ear loops for quick and easy donning
  • Clip connector for secure fastening at back of head available to purchase separately
Providing high particle and bacterial filtration efficiency, the BioClean™ MEA Cleanroom Looped Face Mask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment. The mask features ear loops for quick donning and doffing. (ClipMore Product Information