Anti-DERP2 Human Monoclonal Antibody [clone: H.AK6.A3.F4]

Leverandør: Novus Biologicals
DDX0210P-100
NOVUDDX0210P-100EA 10800 NOK
NOVUDDX0210P-100
Anti-DERP2 Human Monoclonal Antibody [clone: H.AK6.A3.F4]
Antistoffer
The DERP2 Antibody (H.AK6.A3.F4) from Novus Biologicals is a human monoclonal antibody to DERP2. This antibody reacts with human, other. The DERP2 Antibody (H.AK6.A3.F4) has been validated for the following applications: ELISA.

Type: Primary
Antigen: DERP2
Clonality: Monoclonal
Clone: H.AK6.A3.F4
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 Kappa
Reactivity: Human, Other
Bestill nå

SPESIFIKASJONER

Antigensymbol DERP2
Environmentally Preferable
Antigen synonymer Allergen Der p II|Mite group 2 allergen Der p 2|Allergen=Der p 2|DPX|Allergen Der p2|DERP-2
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon H.AK6.A3.F4
Reaktivitet Human
Vert Human
Isotype IgG1 kappa
ELISA Yes
Immunogen Derp-2 was used as immunogen.
Rensing Ion exchange chromatography
Konsentrasjon 0,5 mg/ml
Lagringsbuffer Tris-NaCl (pH 8,0)
Lagringstemp Store at −20 °C. Avoid freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR