Anti-HIF1A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: H1alpha67]

Leverandør: Novus Biologicals
NB100-123PCP
NOVUNB100-123PCPEA 9000 NOK
NOVUNB100-123PCP
Anti-HIF1A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: H1alpha67]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR