Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]

Leverandør: Novus Biologicals
NB100-131 NB100-131SS
NOVUNB100-131EA 8225 NOK
NOVUNB100-131 NOVUNB100-131SS
Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: ESEE122
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR