Anti-EPAS1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ep190b]

Leverandør: Novus Biologicals
NB100-132H
NOVUNB100-132HEA 9875 NOK
NOVUNB100-132H
Anti-EPAS1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ep190b]
Antistoffer
Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: monoclonal
Clone: ep190b
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR