Anti-RAD51L1 Mouse Monoclonal Antibody [clone: 1 H3/13]

Leverandør: Novus Biologicals
NB100-176 NB100-176SS
NOVUNB100-176EA 6825 NOK
NOVUNB100-176 NOVUNB100-176SS
Anti-RAD51L1 Mouse Monoclonal Antibody [clone: 1 H3/13]
Antistoffer
Mouse Monoclonal Rad51L1 Antibody (1 H3/13). Tested Applications: Western Blot. Tested Reactivity: Human, Primate, Mouse (-).

Type: Primary
Antigen: RAD51L1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR