Anti-PDK1 Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
NB100-2383V
NOVUNB100-2383VEA 7825 NOK
NOVUNB100-2383V
Anti-PDK1 Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR