Anti-PDK1 Rabbit Polyclonal Antibody (DyLight® 650)

Leverandør: Novus Biologicals
NB100-2384C
NOVUNB100-2384CEA 7825 NOK
NOVUNB100-2384C
Anti-PDK1 Rabbit Polyclonal Antibody (DyLight® 650)
Antistoffer
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol PDPK1
Antigen-navn 3-phosphoinositide dependent protein kinase 1
Environmentally Preferable
Antigen synonymer 3-phosphoinositide dependent protein kinase 1|Phosphoinositide-dependent kinase-1|PDK1|PDPK2|PDPK2P|PRO0461
Antistofftype Primary
Klonalitet Polyclonal
Konjugering DyLight® 650
Reaktivitet Human
Vert Rabbit
Gen-ID 5163
Isotype
Western blot Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR