Anti-RNF5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LHM 2]

Leverandør: Novus Biologicals
NB100-2688PE
NOVUNB100-2688PEEA 7625 NOK
NOVUNB100-2688PE
Anti-RNF5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LHM 2]
Antistoffer
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR