Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: PyMT]

Leverandør: Novus Biologicals
NB100-2749F
NOVUNB100-2749FEA 7950 NOK
NOVUNB100-2749F
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: PyMT]
Antistoffer
Type: Primary
Antigen: Polyoma Virus, Medium T Antigen
Clonality: monoclonal
Clone: PyMT
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Virus
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR