Anti-CERK Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
NB100-2911B
NOVUNB100-2911BEA 7825 NOK
NOVUNB100-2911B
Anti-CERK Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CERK (ceramide kinase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CERK
Antigen-navn Ceramide kinase
Environmentally Preferable
Antigen synonymer CERK|Acylsphingosine kinase|LK4.|EC 2.7.1.138|Lipid kinase 4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Human
Vert Rabbit
Gen-ID 64781
Isotype
Western blot Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR