Anti-NTHL1 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB100-302 NB100-302SS
NOVUNB100-302EA 6500 NOK
NOVUNB100-302 NOVUNB100-302SS
Anti-NTHL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR