Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]

Leverandør: Novus Biologicals
NB100-306 NB100-306SS
NOVUNB100-306EA 6825 NOK
NOVUNB100-306 NOVUNB100-306SS
Anti-ATM Mouse monoclonal antibody unconjugated [clone: 10H11.E12]
Antistoffer
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR