Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]

Leverandør: Novus Biologicals
NB100-351 NB100-351SS
NOVUNB100-351EA 6500 NOK
NOVUNB100-351 NOVUNB100-351SS
Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]
Antistoffer
Type: Primary
Antigen: RNASEL (ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent))
Clonality: monoclonal
Clone: 2000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR