Anti-Survivin Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB100-56167
NOVUNB100-56167EA 4250 NOK
NOVUNB100-56167
Anti-Survivin Rabbit Polyclonal Antibody
Antistoffer
The Survivin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Survivin. This antibody reacts with human, mouse, rat, gerbil. The Survivin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000

Type: Primary
Antigen: Survivin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Gerbil
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR