Anti-TRAF1 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB100-56170
NOVUNB100-56170EA 4175 NOK
NOVUNB100-56170
Anti-TRAF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRAF1 (TNF receptor-associated factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR