Anti-CVID Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB100-56516
NOVUNB100-56516EA 5150 NOK
NOVUNB100-56516
Anti-CVID Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CVID
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR