Anti-PRMT5 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB100-56641
NOVUNB100-56641EA 5100 NOK
NOVUNB100-56641
Anti-PRMT5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR