Anti-CAV1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 7C8]

Leverandør: Novus Biologicals
NB100-615H
NOVUNB100-615HEA 9450 NOK
NOVUNB100-615H
Anti-CAV1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 7C8]
Antistoffer
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone: 7C8
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR