Anti-PTPRC Rat Monoclonal Antibody (DyLight® 550) [clone: 30-F11]

Leverandør: Novus Biologicals
NB100-77417R
NOVUNB100-77417REA 4775 NOK
NOVUNB100-77417R
Anti-PTPRC Rat Monoclonal Antibody (DyLight® 550) [clone: 30-F11]
Antistoffer
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: 30-F11
Conjugation: DyLight® 550
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR