Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight® 550)

Leverandør: Novus Biologicals
NB110-40763R
NOVUNB110-40763REA 9450 NOK
NOVUNB110-40763R
Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight® 550)
Antistoffer
Type: Primary
Antigen: TRPA1 (transient receptor potential cation channel, subfamily A, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR