Anti-ATP13A2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Novus Biologicals
NB110-41486F
NOVUNB110-41486FEA 8450 NOK
NOVUNB110-41486F
Anti-ATP13A2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: ATP13A2 (ATPase type 13A2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR