Anti-DGAT1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
NB110-41487B
NOVUNB110-41487BEA 8475 NOK
NOVUNB110-41487B
Anti-DGAT1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DGAT1 (diacylglycerol O-acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR