Anti-PLIN5 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB110-60511 NB110-60511SS
NOVUNB110-60511EA 6925 NOK
NOVUNB110-60511 NOVUNB110-60511SS
Anti-PLIN5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PLIN5 (perilipin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR