Anti-BAP1 Mouse Monoclonal Antibody (Biotin) [clone: 1G8]

Leverandør: Novus Biologicals
NB110-60521B
NOVUNB110-60521BEA 8450 NOK
NOVUNB110-60521B
Anti-BAP1 Mouse Monoclonal Antibody (Biotin) [clone: 1G8]
Antistoffer
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR