Anti-APOE Mouse Monoclonal Antibody (DyLight® 650) [clone: WUE-4]

Leverandør: Novus Biologicals
NB110-60531C
NOVUNB110-60531CEA 9450 NOK
NOVUNB110-60531C
Anti-APOE Mouse Monoclonal Antibody (DyLight® 650) [clone: WUE-4]
Antistoffer
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR