Anti-BECN2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
NB110-60984B
NOVUNB110-60984BEA 7825 NOK
NOVUNB110-60984B
Anti-BECN2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: BECN2 (beclin 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR