Anti-CASP11 Rat monoclonal antibody

Leverandør: Novus Biologicals
NB120-10454
NOVUNB120-10454EA 8600 NOK
NOVUNB120-10454
Anti-CASP11 Rat monoclonal antibody
Antistoffer
The Caspase-11 Antibody (17D9) from Novus Biologicals is a rat monoclonal antibody to Caspase-11. This antibody reacts with human, mouse. The Caspase-11 Antibody (17D9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.

Microarray

Type: Primary
Antigen: Caspase 11
Clonality: Monoclonal
Clone: 17D9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR