Anti-VINC Mouse monoclonal antibody

Leverandør: Novus Biologicals
NB120-11193
NOVUNB120-11193EA 6875 NOK
NOVUNB120-11193
Anti-VINC Mouse monoclonal antibody
Antistoffer
The Vinculin Antibody (VIN-11-5) from Novus Biologicals is a mouse monoclonal antibody to Vinculin. This antibody reacts with human, mouse, bovine, chicken. The Vinculin Antibody (VIN-11-5) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Microarray.

Type: Primary
Antigen: Vinculin
Clonality: Monoclonal
Clone: VIN-11-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine, Chicken
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR