Anti-CDKN2A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5-C3-1]

Leverandør: Novus Biologicals
NB200-174PE
NOVUNB200-174PEEA 7625 NOK
NOVUNB200-174PE
Anti-CDKN2A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5-C3-1]
Antistoffer
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: monoclonal
Clone: 5-C3-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol CDKN2A
Antigen-navn Cyclin-dependent kinase inhibitor 2A
Environmentally Preferable
Antigen synonymer ARF-INK4a, Arf, INK4a-ARF, Ink4a/Arf, MTS1, Pctr1, p16, p16(INK4a), p16INK4a, p19, p19ARF
Antistofftype Primary
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Klon 5-C3-1
Reaktivitet Mouse
Vert Rat
Gen-ID 12578
Isotype IgG2b
Flowcytometri Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR