Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 550)

Leverandør: Novus Biologicals
NB21-1053R
NOVUNB21-1053REA 9800 NOK
NOVUNB21-1053R
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 550)
Antistoffer
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR