Anti-GAP43 Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
NB300-143G
NOVUNB300-143GEA 8600 NOK
NOVUNB300-143G
Anti-GAP43 Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
The GAP-43 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to GAP-43. This antibody reacts with human, mouse, rat, porcine, canine, chicken, primate. The GAP-43 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1-2 ?g/ml

Type: Primary
Antigen: GAP-43
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat,More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR