Anti-PECAM1 Rat Monoclonal Antibody (DyLight® 755) [clone: MEC13.3]

Leverandør: Novus Biologicals
NB600-1475IR
NOVUNB600-1475IREA 8325 NOK
NOVUNB600-1475IR
Anti-PECAM1 Rat Monoclonal Antibody (DyLight® 755) [clone: MEC13.3]
Antistoffer
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEC13.3
Conjugation: DyLight® 755
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR