Anti-CD31 Rat Monoclonal Antibody [clone: MEC13.3] (DyLight® 550)

Leverandør: Novus Biologicals
NB600-1475R
NOVUNB600-1475REA 8525 NOK
NOVUNB600-1475R
Anti-CD31 Rat Monoclonal Antibody [clone: MEC13.3] (DyLight® 550)
Antistoffer
Rat Monoclonal CD31/PECAM1 Antibody (MEC13.3) [DyLight 550]. Tested Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactiv

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 0
Conjugation: DyLight® 550
Epitope:
Host: Rat
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR