Membrane protein pituitary tissue lysate (adult normal)
Leverandør: Novus Biologicals
NB820-60579
NOVUNB820-60579EA 16275 NOK
NOVUNB820-60579
Membrane protein pituitary tissue lysate (adult normal)
Celler
Membrane protein pituitary tissue lysate (adult normal).
Tested applications: Western blot, immunoprecipitation.
Tested reactivity: Human