Anti-VIP mouse monoclonal antibody

Leverandør: Novus Biologicals
NBP1-05163
NOVUNBP1-05163EA 9275 NOK
NOVUNBP1-05163
Anti-VIP mouse monoclonal antibody
Antistoffer
The VIP Antibody (2) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to VIP. This antibody reacts with human, rat, porcine, bovine. The VIP Antibody (2) - BSA Free has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VIP
Clonality: Monoclonal
Clone: 2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat, Porcine, Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR