Anti-CCL2 Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
NBP1-07035G
NOVUNBP1-07035GEA 8450 NOK
NOVUNBP1-07035G
Anti-CCL2 Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol MCP1
Antigen-navn Monocyte Chemoattractant Protein 1
Environmentally Preferable
Antigen synonymer monocyte chemoattractant protein-1|small inducible cytokine A2 (monocyte chemotactic protein 1|small inducible cytokine subfamily A (Cys-Cys)|monocyte secretory protein JE|monocyte chemotactic and activating factor|member 2|monocyte chemotactic protein 1|homologous to mouse Sig-je)|small-inducible cytokine A2|monocyte chemoattractant protein 1|C-C motif chemokine 2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering DyLight® 488
Reaktivitet Human
Vert Rabbit
Gen-ID 6347
Isotype
Western blot Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR