Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 650) [clone: HIC1-5F10]

Leverandør: Novus Biologicals
NBP1-18948C
NOVUNBP1-18948CEA 7825 NOK
NOVUNBP1-18948C
Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 650) [clone: HIC1-5F10]
Antistoffer
Type: Primary
Antigen: HPi4
Clonality: monoclonal
Clone: HIC1-5F10
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR