Anti-HPx1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HIC0-3B3]

Leverandør: Novus Biologicals
NBP1-18951H
NOVUNBP1-18951HEA 8450 NOK
NOVUNBP1-18951H
Anti-HPx1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HIC0-3B3]
Antistoffer
Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol HPx1
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering HRP (Horseradish Peroxidase)
Klon HIC0-3B3
Reaktivitet Human
Vert Mouse
Isotype IgG1
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR