HepG2 Whole Cell Cell Lysate (Adult Tumor)

Leverandør: Novus Biologicals

NBP1-42569
NOVUNBP1-42569EA 3650 NOK
NOVUNBP1-42569
HepG2 Whole Cell Cell Lysate (Adult Tumor)
Celler

HepG2 Whole Cell Cell Lysate (Adult Tumor).


  • Source: Human


Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR