Anti-RCAN1 Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
NBP1-46852G
NOVUNBP1-46852GEA 8450 NOK
NOVUNBP1-46852G
Anti-RCAN1 Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
Type: Primary
Antigen: RCAN1 (regulator of calcineurin 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol RCAN1
Environmentally Preferable
Antigen-navn Regulator of calcineurin 1
Antigen synonymer ADAPT78|DSCR1|Regulator of calcineurin 1|RCAN1|DSC1|Calcipressin-1|Myocyte-enriched calcineurin-interacting protein 1|Adapt78|CSP1|Down syndrome critical region protein 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering DyLight® 488
Reaktivitet Human
Vert Rabbit
Gen-ID 1827
Isotype
Western blot Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR