Anti-HDAC6 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
NBP1-78981B
NOVUNBP1-78981BEA 7825 NOK
NOVUNBP1-78981B
Anti-HDAC6 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol HDAC6
Environmentally Preferable
Antigen-navn Histone deacetylase 6
Antigen synonymer JM21|HD6|PPP1R90|CPBHM
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Human
Vert Rabbit
Gen-ID 10013
Isotype
ImmunoFluorescens Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR