Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NBP1-92384
NOVUNBP1-92384EA 0 NOK
NOVUNBP1-92384
Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody
Antistoffer
The SGLT2 / SLC5A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SGLT2 / SLC5A2. This antibody reacts with human. The SGLT2 / SLC5A2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SGLT2/SLC5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR