Anti-TRPM8 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Novus Biologicals
NBP1-97311F
NOVUNBP1-97311FEA 8450 NOK
NOVUNBP1-97311F
Anti-TRPM8 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR