Anti-HO-1 Mouse Monoclonal Antibody [clone: HO-1-1]
Leverandør: Novus Biologicals
NBP1-97507SS
NOVUNBP1-97507SSEA 3600 NOK
NOVUNBP1-97507SS
Anti-HO-1 Mouse Monoclonal Antibody [clone: HO-1-1]
Antistoffer
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information