Anti-MAVS Mouse Monoclonal Antibody [clone: 58N3B6]

Leverandør: Novus Biologicals
NBP2-22380
NOVUNBP2-22380EA 6550 NOK
NOVUNBP2-22380
Anti-MAVS Mouse Monoclonal Antibody [clone: 58N3B6]
Antistoffer
Type: Primary
Antigen: MAVS (mitochondrial antiviral signaling protein)
Clonality: monoclonal
Clone: 58N3B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR